
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:355-74-8
:
Formula:C₆H₆F₈O₂
MDL Number:MFCD00069087
:262.1 g/mol
Boiling Pt:100 °C/3mmHg
Melting Pt:68 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together