Order Entry
Czech Republic
ContactUsLinkComponent
4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole 95+%
Irritant
4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole 95+%
Catalog # APOSPC5331-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole 95+%
Catalog # APOSPC5331-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:157…159 °C
 :195.10 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:18955-88-9
Formula:C₆H₄F₃NO₃
MDL Number:MFCD01941318