
2-Bromo-3-(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:177420-63-2
:
Formula:C₈H₄BrF₃O₂
MDL Number:MFCD08532493
:269.02 g/mol
Melting Pt:143…145 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together