Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-fluoro-3-hydroxybenzoate
Methyl-4-fluoro-3-hydroxybenzoate
Methyl-4-fluoro-3-hydroxybenzoate
APOSPC5221-1G
:
Apollo Scientific
:
Methyl-4-fluoro-3-hydroxybenzoate
APOSPC5221-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
170.14 g/mol
:
89…92 °C
:
GHS07
:
Ambient
:
214822-96-5
:
C₈H₇FO₃
:
MFCD06797457