2,2-Difluoro-1-phenylcyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:156021-07-7
:
Formula:C₁₀H₈F₂O₂
MDL Number:MFCD18434522
:198.17 g/mol
Storage Temperature:Ambient
Frequently Bought Together