Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
Irritant
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
Catalog # APOSPC52069-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-3-fluoro-2-(hydroxymethyl)pyridine 97%
Catalog # APOSPC52069-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.02 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1206968-92-4
Formula:C₆H₅BrFNO
MDL Number:MFCD16607004