Order Entry
Czech Republic
ContactUsLinkComponent
3-(N-Boc-amino)benzotrifluoride ≥95%
3-(N-Boc-amino)benzotrifluoride ≥95%
Catalog # APOSPC520634-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(N-Boc-amino)benzotrifluoride ≥95%
Catalog # APOSPC520634-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:109134-07-8
Formula:C₁₂H₁₄F₃NO₂
MDL Number:MFCD11040633