
3-Hydroxy-5-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:186593-14-6
:
Formula:C₆H₄F₃NO
MDL Number:MFCD10697687
:163.1 g/mol
Melting Pt:159…162 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together