[[4-(Trifluoromethyl)cyclohexyl]methyl]amine(cis- and trans- mixture) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:361393-85-3
:
Formula:C₈H₁₄F₃N
MDL Number:MFCD07777219
:181.20 g/mol
Storage Temperature:Ambient
Frequently Bought Together