Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 95%
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 95%
Catalog # APOSPC520519-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-5-trifluoromethyl-2H-pyrazole-3-carboxylic acid 95%
Catalog # APOSPC520519-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.11 g/mol
Melting Pt:130…132 °C
Storage Temperature:Ambient
CAS Number:128694-63-3
Formula:C₆H₅F₃N₂O₂
MDL Number:MFCD02090853