Order Entry
Czech Republic
ContactUsLinkComponent
3-(1-Bromoethyl)benzotrifluoride 97%
Corrosive
Irritant
3-(1-Bromoethyl)benzotrifluoride 97%
Catalog # APOSPC5176-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-(1-Bromoethyl)benzotrifluoride 97%
Catalog # APOSPC5176-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :253.0600 g/mol
Boiling Pt:98…105 °C/15mmHg
Density:1.48 g/cm³ (25 °C)
Flash Pt:82 °C
UN:3265
ADR:8,III
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:59770-96-6
Formula:C₉H₈BrF₃
MDL Number:MFCD00018050