Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-2-(trifluoromethoxy)aniline
N-Methyl-2-(trifluoromethoxy)aniline
N-Methyl-2-(trifluoromethoxy)aniline
APOSPC5171E-1G
:
Apollo Scientific
:
N-Methyl-2-(trifluoromethoxy)aniline
APOSPC5171E-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.15 g/mol
:
74…76 °C/15mmHg
:
Ambient
:
2810
:
6.1,III
:
GHS02
:
175278-04-3
:
C₈H₈F₃NO
:
MFCD00052334