
2-Methyl-4,4,4-trifluorobutyric acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:99783-23-0
:
Formula:C₅H₇F₃O₂
MDL Number:MFCD00190640
:156.11 g/mol
Boiling Pt:172…175 °C
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together