
4-(Trifluoromethyl)-1-indanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:68755-42-0
:
Formula:C₁₀H₇F₃O
MDL Number:MFCD07772121
:200.16 g/mol
Boiling Pt:67…72 °C/0.1mmHg
Melting Pt:38…42 °C
Flash Pt:>108.3 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together