Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate
Catalog # APOSPC51028-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.14 g/mol
Storage Temperature:Ambient
CAS Number:556112-92-6
Formula:C₉H₆F₄O₂
MDL Number:MFCD06203706