6-[(4-Trifluoromethyl)phenyl]pyridine-2-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:638214-10-5
:
Formula:C₁₃H₈F₃NO
MDL Number:MFCD06410028
:251.21 g/mol
Storage Temperature:Ambient
Frequently Bought Together