6-(4-Fluorophenyl)nicotinic acid
Catalog # APOSPC510071-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:217.2 g/mol
Storage Temperature:Ambient
CAS Number:223127-24-0
Formula:C₁₂H₈FNO₂
MDL Number:MFCD06410388