Silver bis(trifluoromethanesulphonyl)imide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189114-61-2
:
Formula:C₂AgF₆NO₄S₂
MDL Number:MFCD08457644
:388.00 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
Frequently Bought Together