
Methyl-4-fluorobenzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:403-33-8
:
Formula:C₈H₇FO₂
MDL Number:MFCD00017959
:154.14 g/mol
Boiling Pt:90…92 °C/20mmHg
Density:1.192 g/cm³ (25 °C)
Flash Pt:78 °C
EINECS:206-956-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together