
Methyl-3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175137-08-3
:
Formula:C₁₂H₁₀FNO₂S
MDL Number:MFCD00068133
:251.28 g/mol
Melting Pt:153…155 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together