
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20445-31-2
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD00004184
:234.17 g/mol
Boiling Pt:111 °C/1mmHg
Melting Pt:43…45 °C
Density:1.344 g/cm³ (25 °C)
Flash Pt:113 °C
EINECS:243-829-5
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together