
3-Methoxy-5-(trifluoromethyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:871332-97-7
:
Formula:C₈H₈BF₃O₃
MDL Number:MFCD08056362
:219.95 g/mol
Melting Pt:159…162 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together