Order Entry
Czech Republic
ContactUsLinkComponent
DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid 97%
Irritant
DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid 97%
Catalog # APOSPC5044-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
DL-2-[4-(Trifluoromethoxy)phenoxy]propionic acid 97%
Catalog # APOSPC5044-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.18 g/mol
Melting Pt:76…78 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175204-35-0
Formula:C₁₀H₉F₃O₄
MDL Number:MFCD00052353