Sodium-2,6-dinitro-4-(trifluoromethyl)benzenesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54495-25-9
:
Formula:C₇H₃F₃N₂O₇S
MDL Number:MFCD00059068
:338.14 g/mol
Storage Temperature:Ambient
Frequently Bought Together