Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
Corrosive
Irritant
Health
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
Catalog # APOSPC50326-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Health
N,N-Dimethylanilinium tetrakis(pentafluorophenyl)borate 98%
Catalog # APOSPC50326-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :801.23 g/mol
Melting Pt:227 °C
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:118612-00-3
Formula:C₃₂H₁₂BF₂₀N
MDL Number:MFCD12913898