Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-2-(trifluoromethoxy)benzoic acid
4-Bromo-2-(trifluoromethoxy)benzoic acid
Catalog # APOSPC502695-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2-(trifluoromethoxy)benzoic acid
Catalog # APOSPC502695-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :285.0200 g/mol
Storage Temperature:Ambient
CAS Number:509142-48-7
Formula:C₈H₄BrF₃O₃
MDL Number:MFCD12546979