
3'-Chloro-5'-(trifluoromethoxy)acetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886503-42-0
:
Formula:C₉H₆ClF₃O₂
MDL Number:MFCD06660315
:238.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together