
(2-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85952-32-5
:
Formula:C₈H₆ClF₃O₂S
MDL Number:MFCD04972907
Melting Pt:37…39 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together