Order Entry
Czech Republic
ContactUsLinkComponent
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid 98%
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid 98%
Catalog # APOSPC4996-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid 98%
Catalog # APOSPC4996-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:239…241 °C
Storage Temperature:Refrigerator
CAS Number:874219-42-8
Formula:C₁₃H₁₇BFNO₃
MDL Number:MFCD08235059