

2,3-Difluoro-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1215034-77-7
:
Formula:C₆H₂F₅N
MDL Number:MFCD16610846
:183.08 g/mol
Boiling Pt:82…83 °C/400mmHg
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together