
4-(Difluoromethyl)-1,2-difluorobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1214379-64-2
:
Formula:C₇H₄F₄
MDL Number:MFCD14698573
Boiling Pt:62…64 °C/62mmHg
:164.1 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together