3-(Trifluoromethyl)piperidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:768-31-0
:
Formula:C₆H₁₀F₃N
MDL Number:MFCD02183561
:153.15 g/mol
Boiling Pt:74…75 °C/33mmHg
Density:1.154 g/cm³ (25 °C)
Flash Pt:28 °C
Storage Temperature:Refrigerator
Frequently Bought Together