
4'-Iodo-2'-(trifluoromethyl)acetanilide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:97760-98-0
:
Formula:C₉H₇F₃INO
MDL Number:MFCD00052845
Melting Pt:134…136 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together