

2-Fluoro-4-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118078-66-3
:
Formula:C₆H₃F₄N
MDL Number:MFCD09879262
:165.09 g/mol
Boiling Pt:102…106 °C
Density:1.355 g/cm³ (25 °C)
Flash Pt:23.9 °C
Storage Temperature:Ambient
UN:1992
ADR:3+6.1,III
:GHS02
:GHS06
Frequently Bought Together