Order Entry
Czech Republic
ContactUsLinkComponent
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid 97%
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid 97%
Catalog # APOSPC4932-1G
Supplier:  Apollo Scientific
CAS Number:  
2-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]hydrazono}pentanedioic acid 97%
Catalog # APOSPC4932-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :339.66 g/mol
Melting Pt:222…224 °C
Storage Temperature:Ambient
CAS Number:338422-72-3
Formula:C₁₁H₉ClF₃N₃O₄
MDL Number:MFCD00173606