
3-Fluoro-5-(trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-99-6
:
Formula:C₇H₃ClF₄O₂S
MDL Number:MFCD06660226
:262.61 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together