
4-Bromo-1-(difluoromethyl)-2-fluorobenzene 97%
Catalog # APOSPC49172-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:225.01 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:749932-17-0
Formula:C₇H₄BrF₃
MDL Number:MFCD06657963