
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23779-95-5
:
Formula:C₁₁H₆F₃NO₃
MDL Number:MFCD00047603
:GHS07
Storage Temperature:Ambient
Frequently Bought Together