Order Entry
Czech Republic
ContactUsLinkComponent
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid 97%
Irritant
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid 97%
Catalog # APOSPC4869F-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hydroxy-6-(trifluoromethyl)quinoline-3-carboxylic acid 97%
Catalog # APOSPC4869F-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:49713-47-5
Formula:C₁₁H₆F₃NO₃
MDL Number:MFCD00236691