
2-Hydroxy-5-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33252-63-0
:
Formula:C₆H₄F₃NO
MDL Number:MFCD00042315
:163.1 g/mol
Melting Pt:145…149 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together