Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole 97%
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole 97%
Catalog # APOSPC4864-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Hydroxy-n-propyl)-5-(trifluoromethyl)benzimidazole 97%
Catalog # APOSPC4864-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.22 g/mol
Melting Pt:139…141 °C
Storage Temperature:Ambient
CAS Number:175135-15-6
Formula:C₁₁H₁₁F₃N₂O
MDL Number:MFCD00067735