Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-2-fluoro-4-methylbenzonitrile 98%
5-Bromo-2-fluoro-4-methylbenzonitrile 98%
Catalog # APOSPC48632-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2-fluoro-4-methylbenzonitrile 98%
Catalog # APOSPC48632-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.0300 g/mol
Storage Temperature:Refrigerator
CAS Number:1269493-45-9
Formula:C₈H₅BrFN
MDL Number:MFCD18452117