
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33252-64-1
:
Formula:C₆H₃F₃N₂O₃
MDL Number:MFCD00153193
:208.096 g/mol
Melting Pt:189…190 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together