
2-Bromo-1-iodo-4-(trifluoromethoxy)benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:883546-30-3
:
Formula:C₇H₃BrF₃IO
MDL Number:MFCD07369009
:366.9018 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together