1-(4-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSPC48295-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.21 g/mol
Storage Temperature:Ambient
CAS Number:56617-43-7
Formula:C₁₁H₁₀FNO₃
MDL Number:MFCD01569311