
4-Fluoro-3-(trifluoromethyl)cinnamic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:239463-90-2
:
Formula:C₁₀H₆F₄O₂
MDL Number:MFCD00236298
:234.15 g/mol
Melting Pt:141…144 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together