
2-Bromo-1-(4-chloro-2-fluoro-5-methylphenyl)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338982-26-6
:
Formula:C₉H₇BrClFO
MDL Number:MFCD01935937
:265.52 g/mol
Melting Pt:78…81 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together