
1-[(Ammoniooxy)methyl]-2-chloro-4-fluorobenzene chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:317821-68-4
:
Formula:C₇H₈Cl₂FNO
MDL Number:MFCD01114574
Melting Pt:175…180 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together