
2-[(Ammoniooxy)methyl]-1-chloro-3-fluorobenzene chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:93081-15-3
:
Formula:C₇H₈Cl₂FNO
MDL Number:MFCD01114572
Melting Pt:184…188 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together