Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride 95%
Irritant
1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride 95%
Catalog # APOSPC4622-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride 95%
Catalog # APOSPC4622-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:154…156 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:113211-36-2
Formula:C₉H₁₁ClF₃NO₂
MDL Number:MFCD01114594